DescriptionSoftware for CEL-24X Series sound level meters
Warranty3 month
Key Features
Simple 2 button operation
Wide range from 30 to 130dB
A or C frequency weighting
Slow, Fast and Impulse time weightings
Current, highest and average noise level shown in display
Time history trend graphically displayed
USB output port to for simple data logging with pc or download of stored data
Auto calibration at selected decibel level
Scrolling sound level display
¼ inch threaded socket for tripod mounting applications
Model CEL-242 (50537-01)
Offers simple data logging of noise level readings for download to a computer
Model CEL-244 (50537-02)
Adds the time averaging functions for Leq and Lavg for OSHA measurements
Model CEL-246 (50537-03)
Features enhanced averaging version with simple data logging memory
Applications
Industrial hygiene measurements of noise in the workplace
Machinery noise surveys
Assessing hearing protectors using the NRR method
Transportation noise measurements
Building acoustic monitoring
3 month warranty
More About this Item
The CEL-240 series is a completely new range of sound level meters that combine the power of Digital Signal Processing (DSP) technology and simplicity and low cost of a traditional analog instrument. The meter has a low and high measurement range that covers a full 70dB on each setting and include A and C frequency scales and slow, fast and impulse time responses. A non-decaying maximum hold feature on every display captures and displays the highest sound pressure level of any noise until reset by the user. The optional software package is ideal for datalogging in spreadsheet format file for analyzing measurement data further.