Capp ExpellPlus Sterile, Filtered, Pipette Tips, 20 uL; 10 Racks x 96 Tips Each
Representative image only.
Capp Mfr # 5030062C – Item # WZ-25201-92
Accessories and tips for your CAPP® ecopipette™ Single-Channel Variable-Volume Pipettor
  • “Go Green” and “Save Some Green”
  • Compatible with the 25201-06 Ecopipette pipettor
PRODUCT OPTIONS
£91.43GBP / Pkg of 960
£109.72GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)20
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)0.02
  • Max Dispensing (mL)0.02
  • Number Of Channels1
  • Color CodeNatural
  • Package Of10 racks; 96 tips per rack
  • Compatible withCAPP® Ecopipette
  • DescriptionSterile, Filtered, Pipette Tips, 20 uL; 10 Racks x 96 Tips Each
Key Features
  • “Go Green” and “Save Some Green”
  • Compatible with the 25201-06 Ecopipette pipettor
More About this Item
These replacement parts and tips are suitable for use with the CAPP® ecopipette™ Single-Channel Variable-Volume Pipettors, models 25201-02 through -16. Pipettors sold separately.
Special polymers and ultra-smooth inner surfaces virtually eliminate sample loss due to retention or binding. Filters minimize the risk of cross-contamination of samples and the pipette. These sterile tips are also certified free of RNase, DNase, pyrogens, RNA, and DNA.