This item is now discontinued. Please browse similar items
Capp Ecopipette Replament Volume Contol Knob, 2-20 µL, 1/ea
Representative image only.
Capp Mfr # 20-VZ – Item # WZ-25201-33
Accessories and tips for your CAPP® ecopipette™ Single-Channel Variable-Volume Pipettor
  • “Go Green” and “Save Some Green”
  • Compatible with the 25201-06 Ecopipette pipettor
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)20
  • Min Volume (µL)2
  • Number Of Channels1
  • Color CodeGray
  • Compatible withCAPP® Ecopipette
  • DescriptionReplamentVolumeContolKnob,2-20uL;Each
  • Warranty3 year
3 Year Warranty
Key Features
  • “Go Green” and “Save Some Green”
  • Compatible with the 25201-06 Ecopipette pipettor
  • 3 year warranty
More About this Item
These replacement parts and tips are suitable for use with the CAPP® ecopipette™ Single-Channel Variable-Volume Pipettors, models 25201-02 through -16. Pipettors sold separately.
The control knob can be inserted in either direction to allow right or left handed operation. Since the volume display is on the control knob, your fingers do not obscure the display from sight. Volume setting is easy and requires half the number of turns compared to other pipettors. The volume locking device prevents unwanted volume changes.