CappOrigami™ Reagent Reservoir, Polypropylene, Sterile, 40 mL; 5/Bag, 10 Bags/Pk
Representative image only.
Capp Mfr # CA40511 – Item # WZ-25019-66
Innovative design minimizes reagent waste and saves valuable storage space
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • Accommodates 12-channel pipettors
  • 3 year warranty
PRODUCT OPTIONS
£90.60GBP / Pkg of 50
£108.72GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)40
  • Wetted MatterialPolypropylene
  • Package Of5/Bag, 10 Bags/Pk
  • Compatible with12-channel pipettors
  • DescriptionReagent Reservoir, Polypropylene, Sterile, 40 mL; 5/Bag, 10 Bags/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • Accommodates 12-channel pipettors
  • 3 year warranty
More About this Item
CAPP® CappOrigami™ reagent reservoirs are designed to be shipped and stored as flat sheets to save space and reduce shipping costs. The polypropylene construction features a sharp V-shape that makes pipetting easier, prevents the formation of bubbles in the pipette tips, and allows you to use more accurate amounts of reagent and avoid expensive waste. The easy- to-use design features prefolded lines — simply push the sides to ''assemble''. The folds ensure that the reservoirs stay upright.