This item is now discontinued. Please browse similar items
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 18G 2-7/8"
Representative image only.
Cadence Mfr # 7977 – Item # WZ-25701-79
Quality and reliability for all your emulsifying needs
  • Luer lock tip for use with glass syringes
  • Available with or without reinforcing bar
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Lock
  • Needle Gauge18
  • Needle Length (in)2.875
  • MaterialGlass
  • DescriptionMicro-Emulsifying Needle with Reinforcing Bar, 18G 2-7/8\"
  • Warranty6 month
6 Month Warranty
Key Features
  • Luer lock tip for use with glass syringes
  • Available with or without reinforcing bar
  • 6 month warranty
More About this Item
Micro-Emulsifying needles are used in many applications including allergens. Mix two liquids by locking two syringes on the double hub needle and pump fliuids between the two syringes to produce a uniform mixture. Syringes are available with or without a reinforcing bar and in the overall lengths of 1-7/8 inches and 2-7/8 inches. Use with Luer Lock Tip glass syringes.