This item is now discontinued. Please browse similar items
BW Technologies Microdock II GasAlertQuattro Docking Module, and Charger
Representative image only.
BW Technologies Mfr # DOCK2-0-1C1N-00-G – Item # WZ-86419-65
Quick and easy compliance — automatically maintain accurate records
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGasAlertQuattro Docking Module, and Charger
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • 2 year warranty
More About this Item
Manage the calibration and bump testing of your GasAlert portable gas detectors with
these calibration systems. Simplifying the record keeping process, the MicroDock II creates
accurate and user-friendly reports, prints receipts of calibration, sorts and graphs data,
and archives information. Results are stored on the MMC card for easy download to your
computer through the USB interface.
Also features automatic verification of audible and visual alarms. Calibrate multiple units
by adding the appropriate docking modules (up to 10). Connect multiple MicroDock II
systems via a LAN.