Bushing, PTFE, 3/8" NPT(M) X 1/8" NPT(F)
Representative image only.
Item # WZ-34030-44
Maintain a permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
£59.27GBP / Each
£71.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPTFE
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeNPT(M) x NPT(F)
  • Thread size NPT(M) (port 1) (in)3/8
  • Thread size NPT(F) (port 2) (in)1/8
  • DescriptionBushing, PTFE, 3/8" NPT(M) X 1/8" NPT(F)
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.