This item is now discontinued. We recommend the following as suggested replacements: WZ-06286-20
Burkle BeefSteaker 5388-1000 Food core sampler, SS, 25 mm round
Representative image only.
Burkle Mfr # 5388-1000 – Item # WZ-06042-99
Obtain surface samples from solids and semisolids
  • Sturdy 420 stainless steel construction withstands repeated use
  • Vented design prevents air compression while allowing easy specimen retrieval
  • Sterilize by autoclave or flame
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Diameter (in)1
  • Diameter (cm)2.5
  • DescriptionMeat Sampler, SS; 25 mm dia.
  • Warranty15 month
15 Month Warranty
Key Features
  • Sturdy 420 stainless steel construction withstands repeated use
  • Vented design prevents air compression while allowing easy specimen retrieval
  • Sterilize by autoclave or flame
  • 15 month warranty
More About this Item
Obtain bacteriological samples in accordance with HACCP (Hazard Analysis and Critical Control Points)—ideal for slaughterhouses and butcher shops. Works on solids and semisolids for histological, food, textile, and agricultural purposes as well.
Sampler cuts out a 1" (25-mm) diameter specimen from skin or other exposed tissues. Vent hole prevents air entrapment and aids in sample removal.