This item is now discontinued. We recommend the following as suggested replacements: WZ-06264-24
Burkle 5378-3009 Disposable Sampling Spatula, PS, Blue, 150 mm; 10/PK
Representative image only.
Burkle Mfr # 5378-3009 – Item # WZ-06042-75
EU and FDA compliant to safely handle food samples
  • Suitable for food-grade applications
  • Individually packaged in easy-to-open bags
  • Produced and assembled according to cleanroom ISO Class 7
  • Bright blue color reduces chance of being left behind in sample
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)10 2/5
  • Overall Length (cm)26.3
  • MaterialPolystyerene
  • SterileYes
  • Description3009 Disposable Sampling Spatula, PS, FDA Compliant, Blue; 150 mm
  • Warranty15 month
15 Month Warranty
Key Features
  • Suitable for food-grade applications
  • Individually packaged in easy-to-open bags
  • Produced and assembled according to cleanroom ISO Class 7
  • Bright blue color reduces chance of being left behind in sample
  • 15 month warranty
More About this Item
The ergonomically shaped spatulas feature a stable handle and sharp blade allowing it to penetrate directly into paper or plastic sacks—ideal for sampling powders, granulates, and pastes. The spatulas are 10 3/8” (26.3 cm) long with an insertion depth of 6” (15 cm). A transparent sealing sleeve, available on models 06042-40 and 06042-60, securely transports samples and safeguards against possible contamination. Available non-terilized or presterilized via gamma irradiation.