This item is now discontinued. Please browse similar items
Burkle 5317-0030 3 funnel Emptying Device, Stainless Steel; 100 cm
Representative image only.
Burkle Mfr # 5317-0030 – Item # WZ-06041-46
Enhance efficiency and safety of your zone samplers with these accessories
  • Constructed of ultrapure materials for a durable design
  • Accessories are ideal for use with zone samplers for powders and granulates
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material304 Stainless Steel
  • Length (in)39.4
  • Length (cm)100
  • Description3 funnel emptying device, SS,100 cm
  • Warranty15 Months
15 Month Warranty
Key Features
  • Constructed of ultrapure materials for a durable design
  • Accessories are ideal for use with zone samplers for powders and granulates
  • 15 month warranty
More About this Item
The emptying device with 3 funnels (model 06041-46) is made of 304 stainless steel and allows for comfortable and fast emptying of multiple samples directly into a container. The individual single funnel (model 06041-47) can be set to different distances of the collection chambers, and any length of multichamber collection can be emptied. Use a cleaning brush for safe and reliable operation of the samplers—both are made with PVC bristles with a durable stainless-steel handle. Model 06040-92 has a 25 mm diameter while model 06040-93 has a larger 50 mm diameter.