This item is now discontinued. Please browse similar items
Burkle 5309-0301 Cable for Manual Drum Reel, 1.25 mm, SS; 10 m
Representative image only.
Burkle Mfr # 5309-0301 – Item # WZ-06041-23
Safely support your dipping bottles and immersion cylinders
  • 15 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Length (ft)33
  • DescriptionCable for manual drum reel, 1.25 mm, SS; 10 m
  • Warranty15 month
15 Month Warranty
Key Features
  • 15 month warranty
More About this Item
Special manually-operated drum reels with large winding diameters are recommended for safely lowering of Bürkle submersible liquid samplers. Various reels and cables are available to meet your application needs.