This item is now discontinued. Please browse similar items
Bud Industries TR-6102 Enclosure for DIN-Mount Controllers, Medium/Lg Size
Representative image only.
Bud Industries Mfr # TR-6102 – Item # WZ-94503-20
Convert your DIN controllers into a portable desktop instrument
  • Corner gussets provides additional strength
  • Carrying handle may be used as tilting bail
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Outside Width (in)11.06
  • Outside Height (in)7
  • Outside Depth (in)13.25
  • DescriptionEnclosure for DIN-Mount Controllers, Medium/Lg Size
  • Warranty3 Months
3 Month Warranty
Key Features
  • Corner gussets provides additional strength
  • Carrying handle may be used as tilting bail
  • 3 month warranty
More About this Item
The versitality makes it ideal to use as a portable enclosure, a desk top instrument case, or directly into a larger system. Front panel is contructed of 0.09" aluminum and can be easily stamped out to fit controllers and panel meters from 1/16 to 1/2 DIN sizes. Top and bottom panels are constructed from 0.05" aluminum and are easily removed for access to the dust-free interior. Enclosure is finished with a royal blue texture with a white frame. Enclosure comes completely assembled with all hardware included.