This item is now discontinued. Please browse similar items
Bud Industries SNB3730-SS NEMA 4X Stainless Steel Enclosure
Representative image only.
Bud Industries Mfr # SNB-3730-SS – Item # WZ-94502-83
Designed to meet NEMA Type 1, 2, 4, 4X, and IP65 ratings
  • Continuous seam welded 16-gauge steel or stainless steel construction
  • Hinged cover clamps to the base with keyed quarter-turn latch for a watertight seal
  • External mounting brackets so box can be mounted without opening cover
  • Bonding studs ground the base and door
  • Easily machined and modified for specific application
  • Smooth gray powder-coat finish; includes clear zinc plate internal panel and hardware
  • UL listed type 4, 12, and 13
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Outside Width (in)7.87
  • Outside Height (in)9.84
  • Outside Depth (in)5.91
  • DescriptionNEMA 4X Stainless Steel Enclosure
  • Warranty1 Year
1 Year WarrantyIP 65
Key Features
  • Continuous seam welded 16-gauge steel or stainless steel construction
  • Hinged cover clamps to the base with keyed quarter-turn latch for a watertight seal
  • External mounting brackets so box can be mounted without opening cover
  • Bonding studs ground the base and door
  • Easily machined and modified for specific application
  • Smooth gray powder-coat finish; includes clear zinc plate internal panel and hardware
  • UL listed type 4, 12, and 13