This item is now discontinued. Please browse similar items
Bud Industries NG-9511 Cable Gland, PG-7 Mount Thread, 3 to 4.3 mm
Representative image only.
Bud Industries Mfr # NG-9511 – Item # WZ-94501-38
Ideal for Bud Industries NEMA enclosures
  • UL94V-2 flammability rating
  • -40 to 100°C operating temperature range
  • 2 Nm (18"/lb) recommeded torque
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCable Gland, mounting size PG-7
  • Warranty1 Year
1 Year Warranty
Key Features
  • UL94V-2 flammability rating
  • -40 to 100°C operating temperature range
  • 2 Nm (18"/lb) recommeded torque
  • 1 year warranty
More About this Item
The cable glands consists of a black molded nylon body, hexagonal nut, cable nut, a rubber gasket, and cable seal. Cable seal maintains NEMA rating for cable entry or exit.