This item is now discontinued. Please browse similar items
Bud Industries NFL 6630 NEMA 4X Fiberglass Box with Latch Cover and Mounting Brackets
Representative image only.
Bud Industries Mfr # NFL-6630 – Item # WZ-94501-90
Fiberglass-reinforced plastic provides outstanding protection against corrosion
  • Watertight, chemical resistant, durable, and impervious to dents
  • Stainless steel hinge, screws, and wall mounting brackets
  • Includes powder-coated 14-gauge steel removable inner panel
  • Light gray finish
  • NF models have a stainless steel screw-type cover; NFL models have a stainless steel latch-type cover
  • Designed to meet NEMA 1, 2, 4, 4X, 12, 13, and IP65 specifications
  • UL listed and tested to UL508-4X requirements, UL File E194432
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFiberglass
  • Outside Width (in)7.75
  • Outside Height (in)7.75
  • Outside Depth (in)4.8
  • DescriptionNEMA 4X Fiberglass Box with Latch Cover and Mounting Brackets
  • Warranty1 Year
1 Year WarrantyIP 65
Key Features
  • Watertight, chemical resistant, durable, and impervious to dents
  • Stainless steel hinge, screws, and wall mounting brackets
  • Includes powder-coated 14-gauge steel removable inner panel
  • Light gray finish
  • NF models have a stainless steel screw-type cover; NFL models have a stainless steel latch-type cover
  • Designed to meet NEMA 1, 2, 4, 4X, 12, 13, and IP65 specifications
  • UL listed and tested to UL508-4X requirements, UL File E194432