This item is now discontinued. Please browse similar items

Buchi 23012C000 Rotary Evaporator, Dry Ice Condenser, 220V

Representative image only.
Buchi Mfr # 23012C000 – Item # WZ-23012-20
Renowned systems with advanced features for ease of use
  • Quick-action jack makes raising and lowering easy
  • Small footprint takes up minimal lab space
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeDigital (only on bath)
  • Condenser TypeCold-trap
  • Min Speed (rpm)20
  • Max Speed (rpm)280
  • Glass TypeUncoated
  • Joint Size24/40
  • Lift MechanismQuick-action jack
  • Bath Capacity (L)4
  • Min Temperature (° C)20
  • Max Temperature (° C)180
  • Temperature Resolution
  • Temperature Control Accuracy±2°C
  • Vacuum SealPTFE
  • Power (VAC)220
  • Power (watts)1300
  • Power (Hz)50
  • Length (in)21 21/32
  • Width (in)22 41/64
  • Height (in)16 11/32
  • Length (cm)55
  • Width (cm)58
  • Height (cm)42
  • DescriptionRotary Evaporator, Dry Ice Condenser, 220V
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneCanadian Standards Association

Key Features

  • Quick-action jack makes raising and lowering easy
  • Small footprint takes up minimal lab space
  • 1 year warranty

More About this Item

Use these evaporators with flasks from 50 mL to 4 liters. Chemical-resistant PTFE and long-life seals provide practically maintenance-free operation and use. Sparkless, brushless motor provides safe, quiet operation with constant torque from 20 to 280 rpm.

The water/oil bath has a temperature range from 20 to 180°C. Bath has been designed with energy efficiency and safety in mind. Insulating housing prevents contact with hot surfaces; high alloy stainless steel pan is easy to clean. Heating coils are located beneath the surface of the pan to minimize maintenance and simplify cleaning. Use only uncoated flasks when using as an oil bath. Bath features wireless contacts for both power supply and communications, adding to maximum safety for both the unit and user. In the event of a power failure or fault in the bath, the flask is automatically raised to prevent overheating of the product. Each unit features digital display for bath temperature.

Choose from a wide variety of glassware to meet your needs. The diagonal condenser is used for basic distillations and is ideal for where height is limited. The vertical condenser is also used for basic distillations, however takes up less bench space. The vertical assembly also features a connection for a vapor temperature sensor. The cold-trap condenser is ideal for maximizing the condensation due to the low temperatures and no additional cooling water necessary. For distillations needing reflux, choose from either vertical or cold-trap condenser with shutoff. All glassware (except for evaporating flasks) is available with Plastic + Glas coating to minimize breakage and safety concerns.

Note: Direct exposure of Plastic + Glas glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat