This item is now discontinued. Please browse similar items
BTX 45-0100 Petri Dish Electrode for Electroporation, 2 mm gap, 50 mL
Representative image only.
BTX Mfr # 45-0100 – Item # WZ-02894-36
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPetri Dish Electrode for Electroporation, 2 mm gap, 50 mL
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
The Petri Dish Electrodes are designed to be used with a 100mm petri dish which functions as the electroporation chamber. To perform electroporation or electrofusion, the samples are placed in the petri dish and the electrode assembly is lowered into the dish. The electrode assembly is available in 2mm gap sizes. They contain parallel stainless steel electrodes, which produce homogeneous fields. The electrodes are designed to electroporate volumes ranging from 10 to 50 ml.
The 2mm gap Petri Dish Electrode can be used for electroporation of tissues grown right in the petri dish. Simply add the exogenous molecule of interest, mix and electroporate.