BTO Bleed and Temperature Optimized Septa, 3/8" (9.5 mm); 50/Pk
Representative image only.
BTO Mfr # 298705 – Item # WZ-01994-30
Optimized for the most demanding GC and GC/MS applications
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 3 month warranty
PRODUCT OPTIONS
£118.24GBP / Pkg of 50
£141.89GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Description(Bleed/Temp Optimized) Septa, 3/8\" (9.5 mm); 50/Pack
  • Warranty3 month
3 Month Warranty
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 3 month warranty
More About this Item
BTO® (Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.