Branson Sound-Proof Enclosure for Ultrasonic Processors
Representative image only.
Branson Mfr # 101-063-275 – Item # WZ-04715-44
Expand your ultrasonic processing capabilities
PRODUCT OPTIONS
£1,309.21GBP / Each
£1,571.05GBP (incl VAT)
Cole-Parmer Ultrasonic Processor Sound-Abating Enclosure, 30.5
£824.39GBP / Each
£989.27GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithBranson SFX150
  • Height (in)28
  • Width (in)18.5
  • Depth (InchShort)12.5
  • DescriptionSound-Proof Enclosure for Ultrasonic Processors
More About this Item
Get the most out of your Branson ultrasonic processor with these parts and accessories.
Soundproof enclosure reduces mechanical noise by 20 to 25 dB. The cabinet is lined with easy-to-clean waterproof sound-absorbing material, and the fully-transparent door allows process monitoring. Upper and lower openings provide greater access to cup horns or standard horns. Internal column with adjustable clamp plate supports the sample and converter assembly while maintaining a compact footprint. Adapter 04715-45 (order separately) is required for use with the SFX150 ultrasonic processors.