This item is now discontinued. Please browse similar items
Branson Exponential Horn for Sonifiers, 1/2" dia, Solid
Representative image only.
Branson Mfr # 101-147-041 – Item # WZ-04715-56
Customize, upgrade, or replace parts
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (mL)10
  • Max Sample Size (mL)250
  • Tip Diameter (in)0.5
  • DescriptionExponential Horn for Sonifiers, 1/2\" dia, Solid
More About this Item
Ultrasonic processors accommodate a wide variety of horns, chambers, and tips for optimal sample processing in any application.
Horns are fabricated from titanium. Available in different styles and sizes to suit a wide array of applications.