BRAND PD-Tip II Pipette Tips, Non-Sterile Adapter for 25 & 50 mL Tips; 10/PK
Representative image only.
Mfr # 702398 – Item # WZ-24806-14
Fits all repeating pipettes using standard tips
  • High quality virgin plastic construction
  • Encoded for automatic recognition
  • DNA, RNAse, endotoxin, and ATP free
  • Available sterile and non-sterile
  • Adapters can be autoclaved and reused
PRODUCT OPTIONS
£158.36GBP / Pkg of 10
£190.03GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Tip Volume (mL)50
  • Wetted MatterialPolypropylene
  • Package Of10
  • DescriptionPD-Tip™ Non-Sterile Adapter for 25 & 50 mL Tips; 10/pk
Key Features
  • High quality virgin plastic construction
  • Encoded for automatic recognition
  • DNA, RNAse, endotoxin, and ATP free
  • Available sterile and non-sterile
  • Adapters can be autoclaved and reused
More About this Item
PD-Tip™ Precision Dispenser tips are constructed of high-quality virgin high-density polyethylene and polypropylene (0.1 mL tip is polypropylene/LCP). An extended piston minimizes dead volume and maximizes fluid utilization. The precision seal between the piston and cylinder ensures smooth operation, increased accuracy, and increased precision while pipetting. Sterile tips are individually wrapped and certified free of DNA, RNAse, endotoxins, and ATP. 25 and 50 mL tip packs include one autoclavable, polypropylene adapter.
Size automatically recognized by BRAND® HandyStep® touch and electronic, Rainin® AutoRep E and Gilson® Repetman. Compatible with pipettes from BRAND®, Rainin, Gilson (Repetman), Bel-Art and others.