BRAND 388000 Imhoff Sedimentation Cone, SAN, 1000 mL

Representative image only.
BrandTech Mfr # 388000 – Item # WZ-06102-38
Easy drainage with bottom screw on cap
  • 1000 mL volume
  • Graduated per DIN 12672
  • Withstands temperatures up to 85° C
  • 1000 mL Graduated Cone with Screw-On Bottom Cap

PRODUCT OPTIONS

£35.98GBP / Each
£43.18GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • MaterialStyrene Acrylonitrile
  • Sample VolumeGraduated to 1000 mL
  • Cap ConfigurationBottom Screw Cap
  • DescriptionBrandTech Imhoff Sedimentation Cone, SAN, 1000 mL; 1/EA

Key Features

  • 1000 mL volume
  • Graduated per DIN 12672
  • Withstands temperatures up to 85° C
  • 1000 mL Graduated Cone with Screw-On Bottom Cap

More About this Item

This settling cone is made of styrene acrylonitrile to offer exceptional optical clarity for measuring of settled solids.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat