This item is now discontinued. Please browse similar items
Bradley S19-310FW Combination Showers-Shower/eye/facewash station
Representative image only.
Bradley Mfr # S19-310FW – Item # WZ-86000-50
Stay-open valves and flow control ensure constant rinsing flow.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Fixture MaterialChrome-plated brass
  • Bowl MaterialABS plastic
  • Shower Head MaterialABS plastic
  • Supply Pipe MaterialGalvanized steel
  • MountFloor Mount
  • Outlet Connections1-1/4\" NPT(F)
  • Inlet Connection1-1/4\" NPT(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionCombination Showers-Shower/eye/facewash station
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All units feature plastic shower heads with either yellow ABS plastic, corrosive-resistant PVC, or stainless steel eyewash/facewash bowls (10" dia). Each shower comes with safety sign and inspection tag.