This item is now discontinued. Please browse similar items
Bosch TIF8801 Combustible Gas Detector Sensing Tip
Representative image only.
Bosch Mfr # TIF8801 – Item # WZ-10104-73
Optimize performance of your air/gas analyzer
  • Compatible with combustible gas detector 10104-75
  • Excellent for pinpointing the location of combustible gas leaks as low as 5 PPM
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCombustible gas detector 10104-75
  • DescriptionCombustible Gas Detector Sensing Tip
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with combustible gas detector 10104-75
  • Excellent for pinpointing the location of combustible gas leaks as low as 5 PPM
  • 1 year warranty
More About this Item
Replacement sensors, batteries, and other accessories to customize your sampling and analysis system.
Gas detector sold separately.