This item is now discontinued. Please browse similar items
Bio-Works WorkBeads 40 TREN High Capacity IMAC Medium, Tris(2-aminoethyl)amine; 150 mL
Representative image only.
Bio-Works Mfr # KN40603003 – Item # WZ-11947-15
High throughput agarose media for capture of His-tagged proteins
  • Cross-linking method results in a highly porous and physically stable matrix
  • Simple coupling procedure provides fast results
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Immobilized Metal Affinity Chromatography
SPECS & DESCRIPTION
Specifications & Description
  • Media Volume (mL)150
  • Wetted MaterialsTris(2-aminoethyl)amine (TREN) resin
  • ApplicationsImmobilized Metal Affinity Chromatography
  • DescriptionHigh Capacity IMAC Medium, Tris(2-aminoethyl)amine; 150 mL
Key Features
  • Cross-linking method results in a highly porous and physically stable matrix
  • Simple coupling procedure provides fast results
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Immobilized Metal Affinity Chromatography
More About this Item
Separate protein groups on agarose beads with tris(2-aminoethyl)amine (TREN), iminodiacetic acid (IDA), or nitrilotriacetic acid (NTA). Select from uncharged TREN, or charged or uncharged IDA and NTA. Charged media contains IDA or NDA with Ni, Co, Cu, or Zn ions, which are immobilized on the media via the chelating ligand. Metal ion capacity is 50 to 60 µmol/mL.