This item is now discontinued. Please browse similar items
Bio-Simplex DCB10L-2EZ-CF EZ Top Closure Assembly, C-Flex, 2-Port for 10L Carboy
Representative image only.
Bio-Simplex Mfr # DCB10L-2EZ-CF – Item # WZ-62500-47
Optimize performance of your Bio-Simplex Polypropylene Carboys
  • Compatible with the Bio-Simplex Carboys
  • Constructed of durable polypropylene
SPECS & DESCRIPTION
Specifications & Description
  • MaterialC-Flex®
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83 mm
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID9.5 mm
  • DescriptionEZ Top Closure Assembly, C-Flex, 2-Port for 10L Carboy
Key Features
  • Compatible with the Bio-Simplex Carboys
  • Constructed of durable polypropylene
More About this Item
Choose from a wide range of accessories and replacement parts for your Bio-Simplex carboys.
Pre-assembled caps are designed for single use and to withstand temperatures and sterilization to 275°F (135°C). Autoclavable.
Can successfully replace stainless steel closures and eliminate costs for cleaning, validation and assembly. One-piece construction eliminates product loss and contamination due to loose connection. Tubing's smooth inner bore eliminates risk of product entrapment and build-up.
C-Flex® pharmaceutical-grade resin closure has 2 ports and uses 3/8" x 5/8" tubing. Cap includes tubing with standard plug and fitting, flex joint, and 0.2 µm vent filter.