Bio Plas Uni-Capsette 6051 Cassettes, Yellow, 500/pack
Representative image only.
Bio Plas Mfr # 6051 – Item # WZ-48630-01
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Uni-Capsette cassettes have detachable plastic lids to simplify embedding
PRODUCT OPTIONS
£122.14GBP / Pkg of 500
£146.57GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ColorYellow
  • DescriptionCassettes, Yellow, 500/pack
Key Features
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Uni-Capsette cassettes have detachable plastic lids to simplify embedding