This item is now discontinued. Please browse similar items
Bio Plas Capsette 6034 Tissue Processing Cassettes, Blue, 500/pack
Representative image only.
Bio Plas Mfr # 6034 – Item # WZ-48620-04
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlue
  • DescriptionTissue Processing Cassettes, Blue, 500/pack
Key Features
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids