Big Blue Filter cartridge, 20", pleated carbon, 10 micron
Representative image only.
Big Blue Mfr # 155382-43(NCP-20BB) – Item # WZ-29830-33
£52.01GBP / Each
£62.41GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating10
  • Flow Rate10 GPM
  • Filter Length (in)20
  • Capacity20,000 gal at 4 gpm (76,000 L at 15lpm)
  • Max Temperature (° F)150
  • Max Temperature (° C)65.6
  • DescriptionFilter cartridge, 20", pleated carbon, 10 micron
More About this Item
The pleated design offers higher flow rates and dirt-loading capacities than shorter-life block cartridges. The non-cellulose media resists chemical attack and biofilm build-up making the filter ideal for both chlorinated and non-chlorinated water treatment applications. Common applications include post reverse osmosis polishing and well water treatment.