Big Blue Filter cartridge, 10", carbon block, 5 micron
Representative image only.
Big Blue Mfr # 155548-43(EP-BB) – Item # WZ-01512-54
£34.00GBP / Each
£40.80GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating5
  • Flow Rate2 GPM
  • Filter Length (in)10
  • Capacity>22,000 gal at 2gpm (>83,300L at 7.6lpm)
  • Max Temperature (° F)180
  • Max Temperature (° C)82.2
  • DescriptionFilter cartridge, 10", carbon block, 5 micron
More About this Item
The bonded powdered activated carbon filter uses a design to remove organic tastes and odors, chlorine and sediment. Ideal for low or highly variable flow applications since flow cannot channel through the media like granular carbon cartridges††. Cryptosporidium and Giardia cysts are also removed with the 0.5-micron design.
Cartridges are made from FDA compliant materials and meet NSF Standard 42 for materials.
Call Us At
Live chat