This item is now discontinued. Please browse similar items
Bel-Art 42048-8100 Desiccant-Filled Plates; 100-mm diameter; 6/pack
Representative image only.
Bel-Art Mfr # 42048-8100 – Item # WZ-07193-90
  • Silica gel-filled poylystyrene plates
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Plate Size (mm)100
  • Exterior Height (in)0.43
  • Exterior Height (cm)1.1
  • Diameter (InchShort)4
  • Diameter (cm)10
  • MaterialSilica gel-filled Polystyrene
  • DescriptionDesiccant-Filled Plates; 100-mm diameter; 6/pack
  • Warranty6 month
6 Month Warranty
Key Features
  • Silica gel-filled poylystyrene plates
  • 6 month warranty
More About this Item
Replace bulk desiccant with these silica gel-filled plates. The clear polystyrene plates let you visually inspect for color changes from blue to pink as it absorbs moisture. Regenerate desiccant by heating plates in a 122°F (50°C) oven overnight. Plates are individually sealed.