This item is now discontinued. Please browse similar items
Balmac Inc. 191-2 Vibration Transmitter, 0 to In./Sec
Representative image only.
Balmac Inc. Mfr # 191-2 – Item # WZ-98853-15
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Vibration Range - Velocity (in/sec)0
  • Max Vibration Range - Velocity (in/sec)2
  • Max Vibration Range - Velocity (mm/sec)0;51
  • DescriptionVibration Transmitter, 0 to In./Sec
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Solid-state circuitry
  • Two-wire 4-20 mA signal format
  • Same wiring for both power and vibration signal
  • Economical for continuous monitoring of all machines
  • Connects in series with DC power supply and measurement devices
  • Works with single-loop monitors
  • Rugged NEMA 4 weatherproof case
APPLICATIONS
  • Fans, Motors, Pumps and Compressors
  • Controlling Vibrating Conveyors
  • General Purpose Machinery
  • Hazardous Environments