This item is now discontinued. Please browse similar items
B&K Precision XLN10014 1.44kW DC Power Supply 100V/14.4A/USB
Representative image only.
B&K Precision Mfr # XLN10014 – Item # WZ-20035-88
Compact, high density, 1U package provides up to 1.44 kW of power
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Output Voltage0 to 100 VDC
  • Output Current14.4 A
  • Ripple and Noise≤ 8 mVrms, ≤ 80 mVp-p
  • InterfaceUSB
  • Power Rating (watts)1140
  • Description1.44kW DC Power Supply 100V/14.4A/USB
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
These power supplies combine fast average command processing times of less than 50 ms, with the flexibility of industry-standard GPIB, LAN, USB & RS485 interfaces making them ideal for ATE applications. Built-in voltage and current meters display settings and output values concurrently, and units provide standard USB & RS485 interfaces (-GL models include GPIB and LAN). Power supplies feature easy-to-configure master/slave mode for series & parallel connection of up to 4 units, OVP, OCP, OPP protection, and the ability to control up to 31 power supplies from one PC via the RS485 interface.