This item is now discontinued. Please browse similar items
B&K Precision IT-E132 TTL to USB Serial Adapter, Model
Representative image only.
B&K Precision Mfr # IT-E132B – Item # WZ-20033-69
characterize DC power supplies, DC/DC converters, batteries, fuel cells, and solar cells
  • Constant current (CC), resistance (CR), voltage (CV), and power (CP) operation
  • Built-in transient generator with short circuit test
  • Built-in high resolution (0.1 mA/1 mV) voltage and current measurement (model 8500)
  • Over-current, over-voltage, over-power, and over-temperature protection
  • Flexible triggering: Create trigger events by front panel keystroke, back panel TTL signal, or software
  • Remote voltage sensing to compensate for the effect of voltage drop in wires
  • Store up to 25 instrument setups
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTTL to USB Serial Adapter, Model
Key Features
  • Constant current (CC), resistance (CR), voltage (CV), and power (CP) operation
  • Built-in transient generator with short circuit test
  • Built-in high resolution (0.1 mA/1 mV) voltage and current measurement (model 8500)
  • Over-current, over-voltage, over-power, and over-temperature protection
  • Flexible triggering: Create trigger events by front panel keystroke, back panel TTL signal, or software
  • Remote voltage sensing to compensate for the effect of voltage drop in wires
  • Store up to 25 instrument setups