This item is now discontinued. Please browse similar items
B&K Precision DC Electronic Load Module, 1-channel, 500 W/500 V/30 A
Representative image only.
B&K Precision Mfr # MDL505 – Item # WZ-20056-26
Choose up to four load modules for your mainframe
  • Modules and mainframes sold separately
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Depth (in)22.625
  • Power Rating (watts)500
  • DescriptionDC Electronic Load Module, 1-channel, 500 W/500 V/30 A
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Modules and mainframes sold separately
  • 1 year warranty
More About this Item
The MDL Series is a multi-channel modular programmable electronic load system. Seven different modules of programmable DC loads ranging in power from 200 W to 600 W provide users the flexibility to test a wide range of power sources from multi-output AC/DC power supplies to batteries, fuel cells, and photovoltaic arrays.
The mainframe has four slots that can be configured with any assortment of the modules up to 2400 W (up to 4800 W with mainframe extension). The high-performance electronic load modules of the MDL Series are capable of operating in constant current (CC), constant voltage (CV), constant resistance (CR), constant power (CW), and constant impedance (CZ) mode, which uses DSP technology to simulate non-linear loads and realistic loading behavior.