B&K Precision 885 Synthesized LCR/ESR Meter with SMD Probe
Representative image only.
B&K Precision Mfr # 885 – Item # WZ-20020-86
With a built-in direct test fixture, testing the lead components is easy
  • Measurement parameters: Z, L, C, DCR, ESR, D, Q, and F
  • Test conditions: 100Hz, 120Hz, 1kHz, 10kHz, 100KHz (model 886 only), 1 Vrms, 0.25 Vrms, 0.05Vrms
  • 0.5% basic accuracy
£583.00GBP / Each
£699.60GBP (incl VAT)
£147.00GBP / Each
£176.40GBP (incl VAT)
for an additional
£147.00GBP / Each
£176.40GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSynthesized LCR/ESR Meter with SMD Probe
Key Features
  • Measurement parameters: Z, L, C, DCR, ESR, D, Q, and F
  • Test conditions: 100Hz, 120Hz, 1kHz, 10kHz, 100KHz (model 886 only), 1 Vrms, 0.25 Vrms, 0.05Vrms
  • 0.5% basic accuracy
More About this Item
The Model 885 and 886 Synthesized In-Circuit LCR/ESR meters have a wide range of test frequencies up to 10kHz for model 885 and 100kHz for model 886, and has many measurement parameters including Z, L, C, DCR, ESR, D, Q, and F as well. Both are designed for component evaluation on the production line and fundamental impedance testing for bench-top applications. With a built-in direct test fixture, testing the lead components is easy. The optional 4-wire test clip can give a convenient connection to larger components and assemblies with the accuracy of 4-wire testing.