This item is now discontinued. Please browse similar items
B&K Precision 8616 Programmable DC Electronic Load, 500 V/60 A/1200 W Load, 110/220 V
Representative image only.
B&K Precision Mfr # 8616 – Item # WZ-20020-56
Replace space-taking modular load systems with a single versatile unit
  • Test and evaluate a variety of DC sources — power supplies, converters, batteries, battery chargers, and photovoltaic arrays
  • Save up to 100 test setups for quick recall of frequently used protocols
  • Triggers initiated via front panel, external input, timer, or bus
  • Easily integrate into existing systems with USBTMC, RS-232, GPIB, and analog interfaces
  • Included PC software provides front-panel emulation, generation and execution of test sequences, and data logging without needing to write code
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Depth (in)24.5
  • Power Rating (watts)1200
  • Power (VAC)110 / 220
  • Power (Hz)50/60
  • DescriptionProgrammable DC Electronic Load, 500 V/60 A/1200 W Load, 110/220 V
  • Warranty3 Years
3 Year WarrantyConformite Europeenne
Key Features
  • Test and evaluate a variety of DC sources — power supplies, converters, batteries, battery chargers, and photovoltaic arrays
  • Save up to 100 test setups for quick recall of frequently used protocols
  • Triggers initiated via front panel, external input, timer, or bus
  • Easily integrate into existing systems with USBTMC, RS-232, GPIB, and analog interfaces
  • Included PC software provides front-panel emulation, generation and execution of test sequences, and data logging without needing to write code
  • 3 year warranty
More About this Item
The 8600 series programmable DC electronic load has specialized modes for unique applications. The built-in battery test mode measures ampere-hours (Ah), voltage, and capacity level. CR-LED mode simulates the loading behavior of a typical LED. Power supply test modes validate overcurrent and overpower protection functions. Constant current, voltage, resistance, and power modes can be configured for a dynamically changing load with fast switching times. Self-test mode ensures reliability.