B&K Precision 1655A 150 Volt 3 Amp, Isolated Variable AC Power Supply
Representative image only.
B&K Precision Mfr # 1655A – Item # 2001711
  • 1 year warranty
£640.39GBP / Each
£768.47GBP (incl VAT)
£118.89 - £180.34GBP / Each
£142.67 - £216.41GBP (incl VAT)
for an additional
£118.89GBP / Each
£142.67GBP (incl VAT)
for an additional
£180.34GBP / Each
£216.41GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Output Voltage150 VAC
  • Output Current3 A
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
FEATURES:
  • Power line leakage tester
  • 2 Amp continuous output
  • Displays voltage or current readings
  • Isolated transformer to eliminate shock hazard while servicing ''hot'' chassis equipment
  • Compact, rugged