This item is now discontinued. Please browse similar items
Ashcroft D Series Pressure Switch, 15 PSID, 1/4" NPT(F); Single Set Point
Representative image only.
Ashcroft Mfr # D424B15#D – Item # WZ-68361-15
Differential pressure switch designed for your hazardous applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSID)15
  • CaseEpoxy-coated aluminum
  • Process Connection1/4\" NPT(F)
  • Electrical Connections3/4\" NPT(F) conduit
  • OutputSPDT 15A @ 125VAC, 1/2A @ 125VDC
  • Enclosure RatingNEMA 4
  • Media CompatibilityWater-based liquids compatible with Buna-N
  • DescriptionPressure Switch, 15 PSID, 1/4\" NPT(F); Single Set Point
  • Warranty1 year
1 Year WarrantyConformite EuropeenneFM ApprovedIP 66 RatedUL Listed.
Key Features
  • 1 year warranty
More About this Item
Watertight differential pressure switches are ideal for industrial and OEM applications and have fixed set point and deadband. Pressure switche enclosures meet NEMA 3, 4, 4X, 13, and IP66 requirements.
Explosion proof differential pressure switch are designed for hazardous process and industrial applications. Switches have fixed setpoint and deadband. Explosion-proof enclosure meets Div. 1 and 2, MENA 7, 9, and IP66 requirements. These switches meet UL, CSA, ATEX and IECEx approval for hazardous locations.