This item is now discontinued. Please browse similar items
Ashcroft B Series Explosion Proof Pressure Switch, 100 psi, 1/4" NPT(F); Two Set Points
Representative image only.
Ashcroft Mfr # B724B100# – Item # WZ-68360-56
Switches are designed with a fixed set point and deadband
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)100
  • CaseEpoxy-coated aluminum
  • Process Connection1/4\" NPT(F)
  • Electrical Connections3/4\" NPT(F) conduit
  • Enclosure RatingNEMA 7, 9, IP66
  • Media CompatibilityWater-based liquids compatible with Buna-N
  • DescriptionExplosion Proof Pressure Switch, 100 psi, 1/4\" NPT(F); Two Set Points
  • Warranty1 year
1 Year WarrantyConformite EuropeenneFM ApprovedIntrinsically SafeIP 66 RatedUL Listed.
Key Features
  • 1 year warranty
More About this Item
Watertight general purpose pressure switches are ideal for industrial and OEM application.
Explosion proof pressure switches are designed for hazardous process and industrial applications with
ATEX and IECEx approval for hazardous locations.