This item is now discontinued. Please browse similar items
Argos Technologies PolarSafe® Label Strips, Laser Printer, 38 x 6 mm, Multi; 156 x 20 Sheets
Representative image only.
Argos Technologies Item # WZ-04396-78
No need for separate labels—work in cryogenic applications, ultra-low freezers, and incubators
  • Inert polyester with a unique adhesive adheres to glass, plasticware, and most metals
  • No need to self-laminate and sleeker labels fit most storage systems without adding bulk—even cryocanes
  • Strips and dots come in varying sizes, colors, and on rolls for individual labeling or on sheets for use with laser printers
SPECS & DESCRIPTION
Specifications & Description
  • First color changeMulti-Colored
  • Width (in)15/64
  • Length (in)1 1/2
  • DescriptionLabel Strips, Laser Printer, 38 x 6 mm, Multi; 156 x 20 Sheets
Key Features
  • Inert polyester with a unique adhesive adheres to glass, plasticware, and most metals
  • No need to self-laminate and sleeker labels fit most storage systems without adding bulk—even cryocanes
  • Strips and dots come in varying sizes, colors, and on rolls for individual labeling or on sheets for use with laser printers
More About this Item
Argos Technologies PolarSafe labels withstand conventional freezing (-80°C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196°C). Choose the size, color, and format to suit your labeling needs. The wide range of sizes allow you to label your whole experiment permanently. From microcentrifuge tube to cryovial; microtiter plate to storage box.
Ideal for microtiter plates.