This item is now discontinued. Please browse similar items
Argos Technologies Nova® Microcinerator Slide Dryer, 3-Place
Representative image only.
Argos Technologies Item # WZ-04397-13
Enhance the versitility of your Nova® Microcinerator Loop and Needle Sterilizer
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible WithArgos Nova Microincinerator
  • DescriptionMicrocinerator Slide Dryer, 3-Place
More About this Item
The slide dryer is a safe and easy way to use heat already generated by the incinerator to dry slides. This is ideal for dissolving epithelium using KOH mycology technique.
The loop rest protects loop handles from overheating or melting. This accessories fits any loop and attaches easily to the incinerators.
The heating element of your Nova® Microcinerator Loop and Needle Sterilizer will last approximatley 6 months depending on usage. Easily and quikly replace these heating elements to keep your incinerator up and running. Model 04397-12 is for the 120V units and 04397-16 is for the 220V powered incinerators.