Ansell AlphaTec® 58-535B Durable Nitrile Gloves, Size 7; 6 Pair/Bag
Representative image only.
Ansell Mfr # 123819 – Item # WZ-81620-89
Offers superior grip and chemical resistance
  • Safely handle wet or oily substances
  • Reduces chemical leakage onto skin
  • 6 pairs (12 gloves) per polybag
PRODUCT OPTIONS
£35.06GBP / Bag of 6
£42.07GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Size7
  • Length (in)13 45/64
  • Length (cm)34.8
  • ColorGrey and Green
  • GripAnsell Grip™ Technology
  • CuffGauntlet
  • SterileNo
  • Description Durable Nitrile Gloves, Size 7; 6 Pair/Bag
Key Features
  • Safely handle wet or oily substances
  • Reduces chemical leakage onto skin
  • 6 pairs (12 gloves) per polybag
More About this Item
These insulated gloves are manufactured with a polymer coating which does not penetrate the liner, making them liquid proof and chemical resistant. Special grip technology allows you to handle wet or oily objects with less grip force and more control. Ideal for working with chemicals, transferring liquids and solids, handling samples, and warehousing raw materials.