This item is now discontinued. Please browse similar items
Ansell 92-600 Nitrile Gloves, Powder-free, Extra-large
Representative image only.
Ansell Mfr # 92-600 585837 – Item # WZ-81602-75
Standard in comfort, durability, and cleanliness
  • Resist punctures and tear
  • Ambidextrous
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)4
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • ComplianceFDA-accepted
  • SterileNo
  • PowderedNo
  • DescriptionNitrile Gloves, Powder-free, Extra-large
Key Features
  • Resist punctures and tear
  • Ambidextrous
More About this Item
Premium quality gloves are thin enough to allow excellent dexterity, yet tough enough to resist punctures and tears. Gloves are 100% nitrile and contain no fillers, silicones, or plasticizers.