Ansell TouchNTuff® 92-600 Nitrile Gloves, Powder-free, Large
Representative image only.
Ansell Mfr # 92-600 585836 – Item # WZ-81602-65
Standard in comfort, durability, and cleanliness
  • Resist punctures and tear
  • Ambidextrous
PRODUCT OPTIONS
£24.04GBP / Box of 100
£28.85GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)4
  • MaterialNitrile
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • ComplianceFDA-accepted
  • SterileNo
  • PowderedNo
  • DescriptionNitrile Gloves, Powder-free, Large
Key Features
  • Resist punctures and tear
  • Ambidextrous
More About this Item
Premium quality gloves are thin enough to allow excellent dexterity, yet tough enough to resist punctures and tears. Gloves are 100% nitrile and contain no fillers, silicones, or plasticizers.