This item is now discontinued. Please browse similar items
Ansell 37-145 117075 Nitrile Gloves, Unlined, Size 9, 13"L; 12 Pair/Pk
Representative image only.
Ansell Mfr # 37-145 117075 – Item # WZ-09715-30
High-performance nitrile compound provides an outstanding combination of strength and chemical resistance
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)11
  • Size9
  • Length (in)13
  • Length (cm)33
  • ColorGreen
  • LiningNo
  • ComplianceFDA-accepted
  • SterileNo
  • DescriptionNitrile Gloves, Unlined, Size 9, 13\"L; 12 Pair/Pk
Conformite Europeenne
More About this Item
These single-layer gloves are reusable.