This item is now discontinued. Please browse similar items
Ansell 214306 Chemical-Resistant PVA Gloves, 14"L, Size 9, 12 pair/pk
Representative image only.
Ansell Mfr # 214306 – Item # WZ-09715-04
Extreme resistance to strong solvents, including aromatics, aliphatics, and chlorinated solvents
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyvinyl Alcohol (PVA)
  • Size9
  • Length (in)14
  • Length (cm)35
  • ColorRed
  • SterileNo
  • DescriptionChemical-Resistant PVA Gloves, 14\"L, Size 9, 12 pair/pk
More About this Item
Poly vinyl alcohol (PVA) gloves are nearly inert to strong solvents, including aromatics, aliphatics, and chlorinated solvents—chemicals which quickly deteriorate natural rubber, neoprene, and PVC gloves. With PVA gloves, there is little absorption to cause swelling and little extraction to cause shrinking. PVA gloves weigh about half as much as neoprene-coated gauntlets, so they provide a high level of comfort. PVA also offers good resistance to snags, punctures, abrasion, and cuts.
Note: PVA coating is water-soluble. Do not use in water or water-based solutions.