This item is now discontinued. Please browse similar items
Analtech 50511 Uniplates, 20x20cm/Scored/Reversed/250um Thick/15um Particle; 25/Pk
Representative image only.
Analtech Mfr # P50511 – Item # WZ-34102-40
Efficient separations — economically!
  • Constructed of glass backing
SPECS & DESCRIPTION
Specifications & Description
  • Backing MaterialGlass
  • Number of Scored Zones (° C)4
  • Fluorescent Indicator (° C)No
  • Particle size (micron)15
  • Plate Length (cm)20
  • Layer Thickness (micron)250
  • Plate Width (cm)20
  • DescriptionUniplates, 20x20cm/Scored/Reversed/250um Thick/15um Particle; 25/Pk
Key Features
  • Constructed of glass backing
More About this Item
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
Reversed Phase plates use a readily removed nonpolar adsorbent— hydrocarbon-impregnated silica.