This item is now discontinued. Please browse similar items
Analtech 01025 Preparative Uniplates, 10x20cm/Silica G/2000mm Thick/15um Part; 25/Pk
Representative image only.
Analtech Mfr # 01025 – Item # WZ-34103-30
Efficient separations — economically!
  • Constructed of glass backing
SPECS & DESCRIPTION
Specifications & Description
  • MediaCalcium sulfate
  • Backing MaterialGlass
  • Fluorescent Indicator (° C)No
  • Particle size (micron)15
  • Plate Length (cm)10
  • Layer Thickness (mm)2000
  • Plate Width (cm)20
  • DescriptionPreparative Uniplates, 10x20cm/Silica G/2000mm Thick/15um Part; 25/Pk
Key Features
  • Constructed of glass backing
More About this Item
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect preparative plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
Reversed Phase plates use a readily removed nonpolar adsorbent— hydrocarbon-impregnated silica.