This item is now discontinued. Please browse similar items

Amprobe UAT-610 Underground Utility Locator Kit

Representative image only.
Amprobe Mfr # Amprobe UAT-610 – Item # WZ-20034-73
Accurately pinpoint underground utilities and buried services up with accurate depth measurement to 20 feet
  • Multiple tracing modes allow you to locate and trace energized and de-energized utilities in a variety of applications
  • The receiver’s high-contrast display allows for clear viewing in full sunlight and features an automatic backlight for shaded and dark areas
  • Semi-automatic gain control quickly detects tracing signal and allows precise adjustment of the receiver sensitivity
  • Depth measurement up to 20 ft
  • Rated CAT IV 600 V, ensuring safety when working with energized cables
  • Rugged, durable construction: water and dust resistant to IP54 and drop proof to 3.28 ft (1 m)
  • Use the Signal Clamp to induce a signal without making electrical contact (included with the UAT-620)
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Current FrequencyActive tracing: 33 kHz (32,768 Hz) and 8 kHz (8,192 Hz)Passive tracing: 50 / 60 Hz and Radio
  • DisplayReceiver: 4.3 in (109 mm), 320 x 240 BW outdoor LC-Display with auto backlightTransmitter: LCD display (LED backlight) 2.4 in x 1.3 in
  • MeasurementEnergized circuitInduction mode: 33 kHzDirect connection modes: 8 kHz and 33 kHzClamp mode: 8 kHz and 33 kHzDe-Energized circuitInduction mode: 33 kHzDirect connection modes:8 kHz, 33 kHz, A-Lo/A-HiA-Frame: 8 kHzClamp mode: 8 kHz and 33 kHz
  • Safety RatingCAT IV 600 V
  • DescriptionUAT-610 Underground Utility Locator Kit
  • Warranty1 Year
1 Year Warranty

Key Features

  • Multiple tracing modes allow you to locate and trace energized and de-energized utilities in a variety of applications
  • The receiver’s high-contrast display allows for clear viewing in full sunlight and features an automatic backlight for shaded and dark areas
  • Semi-automatic gain control quickly detects tracing signal and allows precise adjustment of the receiver sensitivity
  • Depth measurement up to 20 ft
  • Rated CAT IV 600 V, ensuring safety when working with energized cables
  • Rugged, durable construction: water and dust resistant to IP54 and drop proof to 3.28 ft (1 m)
  • Use the Signal Clamp to induce a signal without making electrical contact (included with the UAT-620)
  • 1 year warranty

More About this Item

Accidentally hitting a utility line during a project can lead to costly repairs and create hazardous public safety situations. Digging in the wrong place can also lead to unnecessary delays and costs for your project, and ultimately, your company. Avoid this disruption with the rugged and durable Amprobe UAT-Series.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat